Tisk…

1-Bromo-3-(trifluoromethylthio)benzene 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Bromophenyl)(trifluoromethyl)sulphane

PC3886-1G PC3886-5G PC3886-25G
APOSPC3886-1GEA 953 CZK
APOSPC3886-1G APOSPC3886-5G APOSPC3886-25G
1-Bromo-3-(trifluoromethylthio)benzene 98%
1-Bromo-3-(trifluoromethylthio)benzene
Vzorec: C₇H₄BrF₃S
MW: 257,0709 g/mol
Bod varu: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00236348
CAS číslo: 2252-45-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR