Tisk…

Propionyl fluoride 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC3875-25G PC3875-5G
APOSPC3875-25GEA 33400 CZK
APOSPC3875-25G APOSPC3875-5G
Propionyl fluoride 97%
Propionyl fluoride
Vzorec: C₃H₅FO
MW: 76,07 g/mol
Bod varu: 42…44 °C/760mmHg
Hustota: 0,947 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039220
CAS číslo: 430-71-7
UN: 3286
ADR: 3+6.1+8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR