5-Fluoro-2-(trifluoromethyl)benzyl alcohol 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC3863-5G
PC3863-1G
APOSPC3863-5GEA
1240
CZK
APOSPC3863-5G
APOSPC3863-1G
5-Fluoro-2-(trifluoromethyl)benzyl alcohol 97%
5-Fluoro-2-(trifluoromethyl)benzyl alcohol
Vzorec:
C₈H₆F₄O MW: 194,13 g/mol Bod tání: 48…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236286 CAS číslo: 238742-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...