Tisk…

2,3-Dibromo-5-(Trifluoromethyl)Pyridine 95%

Dodavatel: Apollo Scientific

PC3861-5G PC3861-1G PC3861-10G
APOSPC3861-5GEA 4570 CZK
APOSPC3861-5G APOSPC3861-1G APOSPC3861-10G
2,3-Dibromo-5-(Trifluoromethyl)Pyridine 95%
2,3-Dibromo-5-(Trifluoromethyl)Pyridine
Vzorec: C₆H₂Br₂F₃N
MW: 304,892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10687233
CAS číslo: 79623-38-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR