Tisk…

3'-(Trifluoromethylthio)acetophenone 94%

Dodavatel: Apollo Scientific
Varování

PC3821-1G PC3821-5G PC3821-25G
APOSPC3821-1GEA 3110 CZK
APOSPC3821-1G APOSPC3821-5G APOSPC3821-25G
3'-(Trifluoromethylthio)acetophenone 94%
3'-(Trifluoromethylthio)acetophenone
Vzorec: C₉H₇F₃OS
Storage Temperature: Refrigerator
MDL Number: MFCD00236331
CAS číslo: 56773-33-2
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR