Tisk…

Oil 10 S

Dodavatel: Apollo Scientific
Varování

PC3815E-1LB PC3815E-4LB
APOSPC3815E-1LBEA 19680 CZK
APOSPC3815E-1LB APOSPC3815E-4LB
Oil 10 S
Oil 10 S
Vzorec: (C₂ClF₃)n
Hustota: 1,868 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS číslo: 9002-83-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR