Oil 10 S
Dodavatel: Apollo Scientific
PC3814A-1LB
PC3814A-5LB
APOSPC3814A-1LBEA
40340
CZK
APOSPC3814A-1LB
APOSPC3814A-5LB
Oil 10 S
Oil 10 S
Vzorec:
(C₂ClF₃)n MW: 800 Hustota: 1.9 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS číslo:
9002-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...