Tisk…

2-Fluoro-5-iodotoluene 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Fluoro-4-iodo-2-methylbenzene, 2-Fluoro-5-iodo-1-methylbenzene

PC3800-25G PC3800-100G PC3800-500G
APOSPC3800-25GEA 622 CZK
APOSPC3800-25G APOSPC3800-100G APOSPC3800-500G
2-Fluoro-5-iodotoluene 99%
2-Fluoro-5-iodotoluene
Vzorec: C₇H₆FI
MW: 236,03 g/mol
Bod varu: 86…87 °C/9mmHg
Storage Temperature: Ambient
MDL Number: MFCD00013710
CAS číslo: 452-68-6
EINECS: 207-206-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR