Tisk…

1-Fluoro-4-iodo-3-nitrobenzene 98%

Dodavatel: Apollo Scientific
Varování

PC3788-1G PC3788-5G
APOSPC3788-1GEA 622 CZK
APOSPC3788-1G APOSPC3788-5G
1-Fluoro-4-iodo-3-nitrobenzene 98%
1-Fluoro-4-iodo-3-nitrobenzene
Vzorec: C₆H₃FINO₂
Bod varu: 110 °C/4mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00153169
CAS číslo: 364-77-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR