1-Fluoro-4-iodobenzene 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Fluoroiodobenzene
PC3780-100G
PC3780-500G
PC3780-50G
APOSPC3780-100GEA
1040
CZK
APOSPC3780-100G
APOSPC3780-500G
APOSPC3780-50G
1-Fluoro-4-iodobenzene 99%
1-Fluoro-4-iodobenzene
Tet. Lett., 33 7499 (1992): Using LDA regioselective metallation can be achieved ortho- to the fluorine atom.J.Med.Chem,40,717(1997):5-iodoanthranilic acid & deriv.
Vzorec:
C₆H₄FI MW: 222 g/mol Bod varu: 182…184 °C Bod tání: –20 °C Hustota: 1,925 g/cm³ (25 °C) Bod vzplanutí: 68 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001052 CAS číslo: 352-34-1 EINECS: 206-522-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...