Bis(hexafluoroacetylacetonato)manganese(II) hydrate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Hexafluoroacetylacetono Manganese(II) Salt, Manganese(II) Hexafluoroacetylacetonate
PC3766-1G
PC3766-5G
PC3766-25G
APOSPC3766-1GEA
2900
CZK
APOSPC3766-1G
APOSPC3766-5G
APOSPC3766-25G
Bis(hexafluoroacetylacetonato)manganese(II) hydrate 97%
Bis(hexafluoroacetylacetonato)manganese(II) hydrate
Vzorec:
C₁₀H₂F₁₂MnO₄ MW: 469,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00042513 CAS číslo: 19648-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...