5-Fluoroindole-2-carboxylic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Fluoro-1H-indole-2-carboxylic acid
PC3758E-1G
PC3758E-25G
PC3758E-5G
APOSPC3758E-1GEA
1040
CZK
APOSPC3758E-1G
APOSPC3758E-25G
APOSPC3758E-5G
5-Fluoroindole-2-carboxylic acid 99%
5-Fluoroindole-2-carboxylic acid
Vzorec:
C₉H₆FNO₂ MW: 179.15 Bod tání: 254…259 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005612 CAS číslo: 399-76-8 EINECS: 206-919-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...