Tisk…

6-Fluorosalicylic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC3756B-1G PC3756B-25G PC3756B-5G
APOSPC3756B-1GEA 622 CZK
APOSPC3756B-1G APOSPC3756B-25G APOSPC3756B-5G
6-Fluorosalicylic acid 97%
6-Fluorosalicylic acid
Vzorec: C₇H₅FO₃
Bod tání: 159…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00052179
CAS číslo: 67531-86-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR