Tetrafluoropyrimidine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,4,5,6-Tetrafluoropyrimidine
PC3755-1G
PC3755-5G
APOSPC3755-1GEA
11540
CZK
APOSPC3755-1G
APOSPC3755-5G
Tetrafluoropyrimidine 97%
Tetrafluoropyrimidine
Vzorec:
C₄F₄N₂ Bod varu: 83…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD10687402 CAS číslo: 767-79-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...