Tisk…

Allyl 1H,1H-heptafluorobutyl ether 97%

Dodavatel: Apollo Scientific

PC3736-25G PC3736-5G
APOSPC3736-25GEA 5820 CZK
APOSPC3736-25G APOSPC3736-5G
Allyl 1H,1H-heptafluorobutyl ether 97%
Allyl 1H,1H-heptafluorobutyl ether
Vzorec: C₇H₇F₇O
Bod varu: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD01862010
CAS číslo: 648-42-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR