Tisk…

Ethyl-5-(trifluoromethyl)picolinate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Ethyl 5-(trifluoromethyl)-2-pyridinecarboxylate

PC3723-250MG
APOSPC3723-250MGEA 1990 CZK
APOSPC3723-250MG
Ethyl-5-(trifluoromethyl)picolinate
Ethyl-5-(trifluoromethyl)picolinate
Vzorec: C₉H₈F₃NO₂
MW: 219,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100197
CAS číslo: 128072-94-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR