3-[(Trifluoromethyl)thio]aniline 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC3714-5G
PC3714-25G
APOSPC3714-5GEA
4570
CZK
APOSPC3714-5G
APOSPC3714-25G
3-[(Trifluoromethyl)thio]aniline 97%
3-[(Trifluoromethyl)thio]aniline
Vzorec:
C₇H₆F₃NS MW: 193.19 g/mol Bod varu: 221…224 °C/11mmHg Hustota: 1.348 g/cm³ (25 °C) Bod vzplanutí: 103 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00190130 CAS číslo: 369-68-6 EINECS: 206-744-4 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...