Tisk…

2,6-Dichloro-4-(trifluoromethoxy)phenylacetic acid 97%

Dodavatel: Apollo Scientific

PC3698-1G PC3698-5G
APOSPC3698-1GEA 4570 CZK
APOSPC3698-1G APOSPC3698-5G
2,6-Dichloro-4-(trifluoromethoxy)phenylacetic acid 97%
2,6-Dichloro-4-(trifluoromethoxy)phenylacetic acid
Vzorec: C₉H₅Cl₂F₃O₃
Storage Temperature: Ambient
CAS číslo: 886503-16-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR