α-Bromo-2-fluorotoulene 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Fluorobenzyl bromide, α-Bromo-o-fluorotoulene
PC3693-100G
PC3693-250G
PC3693-25G
APOSPC3693-100GEA
1240
CZK
APOSPC3693-100G
APOSPC3693-250G
APOSPC3693-25G
α-Bromo-2-fluorotoulene 97%
α-Bromo-2-fluorotoulene
Vzorec:
C₇H₆BrF MW: 189.03 Bod varu: 62…64 °C/15mmHg Hustota: 1.567 Bod vzplanutí: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000324 CAS číslo: 446-48-0 EINECS: 207-169-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...