Tisk…

4,6-Difluorosalicylaldehyde

Dodavatel: Apollo Scientific

Synonyms: 2,4-Difluoro-6-hydroxybenzaldehyde

PC3686-1G PC3686-5G
APOSPC3686-1GEA 2280 CZK
APOSPC3686-1G APOSPC3686-5G
4,6-Difluorosalicylaldehyde
4,6-Difluorosalicylaldehyde
Vzorec: C₇H₄F₂O₂
MW: 158,1023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701048
CAS číslo: 136516-64-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR