Ethyl-2-(4-(trifluoromethyl)phenyl)acetate 96%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 2-(4-(trifluoromethyl)phenyl)acetate, Ethyl 4-(Trifluoromethyl)phenylacetate, 4-(Trifluoromethyl)phenylacetic acid ethyl ester
PC3679-25G
APOSPC3679-25GEA
4270
CZK
APOSPC3679-25G
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate 96%
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate
Vzorec:
C₁₁H₁₁F₃O₂ MW: 232,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07779351 CAS číslo: 721-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...