Tisk…

4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde

Dodavatel: Apollo Scientific

PC3666-1G PC3666-5G
APOSPC3666-1GEA 3910 CZK
APOSPC3666-1G APOSPC3666-5G
4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde
4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde
Vzorec: C₁₃H₈F₃NO₂
MW: 267.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10703517
CAS číslo: 103962-21-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR