Tisk…

3-(4-Bromo-3-fluorophenyl)-1-propene

Dodavatel: Apollo Scientific

Synonyms: 4-Allyl-2-fluorobromobenzene

PC3642-1G PC3642-5G
APOSPC3642-1GEA 6820 CZK
APOSPC3642-1G APOSPC3642-5G
3-(4-Bromo-3-fluorophenyl)-1-propene
3-(4-Bromo-3-fluorophenyl)-1-propene
Vzorec: C₉H₈BrF
MW: 215,07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09801083
CAS číslo: 951888-38-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR