Tisk…

4-(4-Fluorobenzoyl)piperidine hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Fluorophenyl)(piperidin-4-yl)methanone hydrochloride

PC3634-25G
APOSPC3634-25GEA 0 CZK
APOSPC3634-25G
4-(4-Fluorobenzoyl)piperidine hydrochloride 97%
4-(4-Fluorobenzoyl)piperidine hydrochloride
Vzorec: C₁₂H₁₅ClFNO
MW: 243,71 g/mol
Bod tání: 222…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00044912
CAS číslo: 25519-78-2
EINECS: 247-070-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR