4-Fluoro-3-hydroxybenzeneboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Fluoro-3-hydroxyphenylboronic acid, (4-Fluoro-3-hydroxyphenyl)boronic acid
PC3555-1G
PC3555-250MG
PC3555-5G
APOSPC3555-1GEA
2400
CZK
APOSPC3555-1G
APOSPC3555-250MG
APOSPC3555-5G
4-Fluoro-3-hydroxybenzeneboronic acid 98%
4-Fluoro-3-hydroxybenzeneboronic acid
Vzorec:
C₆H₆BFO₃ MW: 155.92 g/mol Bod tání: 264…266 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689483 CAS číslo: 913835-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...