Ethyl 3,5-bis(trifluoromethyl)benzoate
Dodavatel: Apollo Scientific
PC3553-1G
PC3553-5G
APOSPC3553-1GEA
2170
CZK
APOSPC3553-1G
APOSPC3553-5G
Ethyl 3,5-bis(trifluoromethyl)benzoate
Ethyl-3,5-bis(trifluoromethyl)benzoate
Vzorec:
C₁₁H₈F₆O₂ MW: 286.17 g/mol Bod varu: 84 °C/10mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01320684 CAS číslo: 96617-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...