N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine
Dodavatel: Apollo Scientific
Synonyms:
Boc-Phe(4-F)-OH, N-Boc-4-fluoro-L-phenylalanine
PC3551-25G
PC3551-5G
APOSPC3551-25GEA
7440
CZK
APOSPC3551-25G
APOSPC3551-5G
N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine
N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine
Vzorec:
C₁₄H₁₈FNO₄ MW: 283.30 g/mol Bod tání: 83…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079672 CAS číslo: 41153-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...