Tisk…

3-Fluoro-4-pyridineboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Fluoropyridin-4-yl)boronic acid

PC3533-1G PC3533-500MG PC3533-250MG
APOSPC3533-1GEA 1740 CZK
APOSPC3533-1G APOSPC3533-500MG APOSPC3533-250MG
3-Fluoro-4-pyridineboronic acid 97%
3-Fluoro-4-pyridineboronic acid
Vzorec: C₅H₅BFNO₂
MW: 140,91 g/mol
Bod tání: 237…239 °C
Storage Temperature: Ambient
MDL Number: MFCD03788558
CAS číslo: 458532-97-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR