Tisk…

Tris(hexafluoroisopropyl)borate 95%

Dodavatel: Apollo Scientific
Varování

PC3486-1G PC3486-25G PC3486-5G
APOSPC3486-1GEA 3120 CZK
APOSPC3486-1G APOSPC3486-25G APOSPC3486-5G
Tris(hexafluoroisopropyl)borate 95%
Tris(hexafluoroisopropyl)borate
Vzorec: C₉H₃BF₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD00191615
CAS číslo: 6919-80-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR