Undecafluorohexanoic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Perfluorohexanoic Acid, IPC-PFFA-6
PC3454-1G
PC3454-5G
PC3454-10G
APOSPC3454-1GEA
622
CZK
APOSPC3454-1G
APOSPC3454-5G
APOSPC3454-10G
Undecafluorohexanoic acid
Undecafluorohexanoic acid
Vzorec:
C₆HF₁₁O₂ MW: 314.05 g/mol Bod varu: 157 °C Hustota: 1.762 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00198040 CAS číslo: 307-24-4 EINECS: 206-196-6 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...