4-Bromo-1-(2-fluorophenyl)-1H-pyrazole
Dodavatel: Apollo Scientific
PC3444-1G
PC3444-5G
APOSPC3444-1GEA
6120
CZK
APOSPC3444-1G
APOSPC3444-5G
4-Bromo-1-(2-fluorophenyl)-1H-pyrazole
4-Bromo-1-(2-fluorophenyl)-1H-pyrazole
Vzorec:
C₉H₆BrFN₂ Storage Temperature: Ambient |
MDL Number:
MFCD09878403 CAS číslo: 957062-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...