Tisk…

5-Fluoro-2-(methylcarbamoyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (5-Fluoro-2-(methylcarbamoyl)phenyl)boronic acid

PC3436-1G PC3436-250MG
APOSPC3436-1GEA 8100 CZK
APOSPC3436-1G APOSPC3436-250MG
5-Fluoro-2-(methylcarbamoyl)phenylboronic acid
5-Fluoro-2-(methylcarbamoyl)phenylboronic acid
Vzorec: C₈H₉BFNO₃
MW: 196,9700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878343
CAS číslo: 957062-96-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR