3-Methoxy-4-(difluoromethoxy)benzaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(Difluoromethoxy)-3-methoxybenzaldehyde
PC3425-5G
PC3425-1G
APOSPC3425-5GEA
662
CZK
APOSPC3425-5G
APOSPC3425-1G
3-Methoxy-4-(difluoromethoxy)benzaldehyde 97%
3-Methoxy-4-(difluoromethoxy)benzaldehyde
Vzorec:
C9H8F2O3 MW: 202,1548 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03412217 CAS číslo: 162401-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...