Tisk…
2,2,3,3,4,4-Hexafluoro-1,5-pentyl dimethacrylate 97%
Dodavatel: Apollo Scientific
PC3418-1G PC3418-5G
APOSPC3418-1GEA 3940 CZK
APOSPC3418-1G APOSPC3418-5G
2,2,3,3,4,4-Hexafluoro-1,5-pentyl dimethacrylate 97%
2,2,3,3,4,4-Hexafluoro-1,5-pentyl dimethacrylate
Vzorec: C₁₃H₁₄F₆O₄
MW: 348,24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00069086
CAS číslo: 918-36-5