1,2-Diamino-3,4-difluorobenzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,4-Difluoro-1,2-phenylenediamine, 3,4-Difluorobenzene-1,2-diamine, 3,4-Difluoro-o-phenylenediamine
PC3416-1G
PC3416-5G
APOSPC3416-1GEA
622
CZK
APOSPC3416-1G
APOSPC3416-5G
1,2-Diamino-3,4-difluorobenzene 97%
1,2-Diamino-3,4-difluorobenzene
Vzorec:
C₆H₆F₂N₂ Bod tání: 58…60 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00153128 CAS číslo: 153505-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...