1,1,1-Trifluoroacetone oxime 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC3386-25G
PC3386-5G
PC3386-1G
APOSPC3386-25GEA
17660
CZK
APOSPC3386-25G
APOSPC3386-5G
APOSPC3386-1G
1,1,1-Trifluoroacetone oxime 97%
1,1,1-Trifluoroacetone oxime
Vzorec:
C₃H₄F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD06248847 CAS číslo: 431-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...