Tisk…

Sodium iodofluoroacetate 98%

Dodavatel: Apollo Scientific
Varování

PC3377-1G PC3377-5G
APOSPC3377-1GEA 6720 CZK
APOSPC3377-1G APOSPC3377-5G
Sodium iodofluoroacetate 98%
Sodium iodofluoroacetate
Vzorec: C₂HFINaO₂
Storage Temperature: Ambient
MDL Number: MFCD04038357
CAS číslo: 1177620-76-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR