Sodium chlorofluoroacetate 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC3376-1G
PC3376-5G
APOSPC3376-1GEA
2070
CZK
APOSPC3376-1G
APOSPC3376-5G
Sodium chlorofluoroacetate 98%
Sodium chlorofluoroacetate
Vzorec:
C₂HClFNaO₂ Storage Temperature: Ambient |
MDL Number:
MFCD01861160 CAS číslo: 70395-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...