Sodium bromodifluoroacetate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Bromodifluoroacetic Acid Sodium Salt
PC3374-25G
PC3374-5G
PC3374-1G
APOSPC3374-25GEA
1450
CZK
APOSPC3374-25G
APOSPC3374-5G
APOSPC3374-1G
Sodium bromodifluoroacetate 97%
Sodium bromodifluoroacetate
Vzorec:
C₂HBrF₂O₂ MW: 196,91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038355 CAS číslo: 84349-27-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...