Potassium perfluoro-(2-ethoxyethane)sulphonate
Dodavatel: Apollo Scientific
PC3371-25G
PC3371-5G
APOSPC3371-25GEA
19080
CZK
APOSPC3371-25G
APOSPC3371-5G
Potassium perfluoro-(2-ethoxyethane)sulphonate
Potassium perfluoro-(2-ethoxyethane)sulphonate
Vzorec:
C₄F₉KO₄S Storage Temperature: Ambient |
MDL Number:
MFCD00155953 CAS číslo: 117205-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...