Tisk…

2-Chloro-5-(trifluoromethyl)benzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Bromomethyl)-2-chloro-5-(trifluoromethyl)benzene, 2-(Bromomethyl)-1-chloro-4-(trifluoromethyl)benzene

PC3355-1G PC3355-25G PC3355-5G
APOSPC3355-1GEA 622 CZK
APOSPC3355-1G APOSPC3355-25G APOSPC3355-5G
2-Chloro-5-(trifluoromethyl)benzyl bromide 98%
2-Chloro-5-(trifluoromethyl)benzyl bromide
Vzorec: C₈H₅BrClF₃
MW: 273.48
Bod varu: 80 °C/3mmHg
Bod tání: 51
Storage Temperature: Ambient
MDL Number: MFCD00236216
CAS číslo: 237761-77-2
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR