Tisk…

4-Bromo-N,N-dimethyl-2-fluorobenzamide 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-2-fluoro-N,N-dimethylbenzamide

PC3354-1G PC3354-5G
APOSPC3354-1GEA 2690 CZK
APOSPC3354-1G APOSPC3354-5G
4-Bromo-N,N-dimethyl-2-fluorobenzamide 98%
4-Bromo-N,N-dimethyl-2-fluorobenzamide
Vzorec: C₉H₉BrFNO
MW: 246,0763 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878360
CAS číslo: 749927-80-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR