Perfluorohexene-2 80%
Dodavatel: Apollo Scientific
PC3343-5G
PC3343-1G
APOSPC3343-5GEA
7940
CZK
APOSPC3343-5G
APOSPC3343-1G
Perfluorohexene-2 80%
Perfluorohexene-2
Vzorec:
C₆F₁₂ MW: 300.05 g/mol Bod varu: 48…49 °C Hustota: 1.6 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00443180 CAS číslo: 1584-00-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...