4'-Fluoroacetanilide 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Fluoroacetanilide, N-(4-Fluorophenyl)acetamide
PC3340-25G
APOSPC3340-25GEA
1330
CZK
APOSPC3340-25G
4'-Fluoroacetanilide 98%
4'-Fluoroacetanilide
Vzorec:
C₈H₈FNO MW: 153,16 g/mol Bod tání: 151…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016335 CAS číslo: 351-83-7 EINECS: 206-515-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...