Tisk…

Perfluorooctene-2

Dodavatel: Apollo Scientific

PC3332-1G PC3332-25G PC3332-5G
APOSPC3332-1GEA 4570 CZK
APOSPC3332-1G APOSPC3332-25G APOSPC3332-5G
Perfluorooctene-2
Perfluorooctene-2
Vzorec: C₈F₁₆
MW: 400.07 g/mol
Bod varu: 95…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038344
CAS číslo: 65500-50-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR