Tisk…

PTFE

Dodavatel: Apollo Scientific
Varování

Synonyms: Polytetrafluoroethylene, Teflon

PC3329-5KG PC3329-5G
APOSPC3329-5KGEA 0 CZK
APOSPC3329-5KG APOSPC3329-5G
PTFE
PTFE

60 wt % dispersion in water; 0.05-0.5 micron particles.

Vzorec: (CF₂CF₂)n
Hustota: 1,5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00084455
CAS číslo: 9002-84-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR