1-Chloro-3-(trifluoromethoxy)benzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC3328-10G
PC3328-50G
PC3328-5G
APOSPC3328-10GEA
953
CZK
APOSPC3328-10G
APOSPC3328-50G
APOSPC3328-5G
1-Chloro-3-(trifluoromethoxy)benzene 97%
1-Chloro-3-(trifluoromethoxy)benzene
Vzorec:
C₇H₄ClF₃O Bod varu: 140 °C Hustota: 1,37 g/cm³ (25 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00276971 CAS číslo: 772-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...