Tisk…

Tris(perfluorobutyl)amine

Dodavatel: Apollo Scientific
Varování

Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

PC3312C-100G PC3312C-1KG PC3312C-250G
APOSPC3312C-100GEA 6020 CZK
APOSPC3312C-100G APOSPC3312C-1KG APOSPC3312C-250G
Tris(perfluorobutyl)amine
Tris(perfluorobutyl)amine

(Fluorinert is a registered trademark of 3M)

Vzorec: C₁₂F₂₇N
MW: 671,1 g/mol
Bod varu: 178 °C
Bod tání: –52 °C
Hustota: 1,88 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000436
CAS číslo: 311-89-7
EINECS: 206-223-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR