3,5-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxypropyl)iodobenzene 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,2'-(5-Iodobenzene-1,3-diyl)bis(1,1,1,3,3,3-hexafluoropropan-2-ol)
PC3311-5G
PC3311-1G
APOSPC3311-5GEA
11540
CZK
APOSPC3311-5G
APOSPC3311-1G
3,5-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxypropyl)iodobenzene 97%
3,5-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxypropyl)iodobenzene
Vzorec:
C₁₂H₅F₁₂IO₂ MW: 536,06 g/mol Bod varu: 80…81 °C/0,1mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD04038221 CAS číslo: 53173-72-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...