Ethyl trifluoropyruvate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate
PC3288G-100G
PC3288G-25G
APOSPC3288G-100GEA
1870
CZK
APOSPC3288G-100G
APOSPC3288G-25G
Ethyl trifluoropyruvate 98%
Ethyl trifluoropyruvate
Useful prochiral intermediate
Vzorec:
C₅H₅F₃O₃ MW: 170,09 g/mol Bod varu: 95…98 °C Hustota: 1,283 g/cm³ (25 °C) Bod vzplanutí: 31 °C Storage Temperature: Ambient |
MDL Number:
MFCD00114935 CAS číslo: 13081-18-0 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...